Assay Details
| Gene Symbol: tmem30aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212776.2 | NP_997941.2 | 3 - 4 | 659 | 70 | |
| GenBank mRNA | AY391412.1 | - | 3 - 4 | 613 | 70 | |
| BC074040.1 | - | 3 - 4 | 592 | 70 | ||
| BC164201.1 | - | 3 - 4 | 504 | 70 | ||
| EH584307.1 | - | 3 - 4 | 713 | 70 | ||
More Information
Set Membership: |
|

tmem30aa
Genomic Map