Assay Details
| Gene Symbol: gapdhs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213094.2 | NP_998259.1 | 9 - 10 | 896 | 73 | |
| GenBank mRNA | AY818346.1 | - | 8 - 9 | 868 | 73 | |
| BC154822.1 | - | 9 - 10 | 896 | 73 | ||
More Information
Set Membership: |
|

gapdhs
Genomic Map