Assay Details
| Gene Symbol: yy1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213120.1 | NP_998285.1 | 1 - 2 | 603 | 82 | |
| XM_005170229.4 | XP_005170286.1 | 1 - 2 | 841 | 82 | ||
| GenBank mRNA | BC053307.1 | - | 1 - 2 | 603 | 82 | |
| BC155321.1 | - | 1 - 2 | 619 | 82 | ||
More Information
Set Membership: |
|

yy1b
Genomic Map