Assay Details
| Gene Symbol: marcksl1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213133.1 | NP_998298.1 | 1 - 2 | 260 | 74 | |
| GenBank mRNA | BC058848.1 | - | 1 - 2 | 260 | 74 | |
| JF894245.1 | - | 1 - 2 | 179 | 74 | ||
More Information
Set Membership: |
|

marcksl1a
Genomic Map