Assay Details
| Gene Symbol: agxtb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213162.1 | NP_998327.1 | 10 - 11 | 1171 | 79 | |
| GenBank mRNA | BC056520.1 | - | 10 - 11 | 1171 | 79 | |
More Information
Set Membership: |
|

agxtb
Genomic Map