Assay Details
| Gene Symbol: agxtb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213162.1 | NP_998327.1 | 3 - 4 | 519 | 97 | |
| GenBank mRNA | BC056520.1 | - | 3 - 4 | 519 | 97 | |
| BC067638.1 | - | 3 - 4 | 505 | 97 | ||
More Information
Set Membership: |
|

agxtb
Genomic Map