Assay Details
| Gene Symbol: stk25b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213308.1 | NP_998473.1 | 7 - 8 | 1216 | 70 | |
| GenBank mRNA | BC045867.1 | - | 7 - 8 | 1192 | 70 | |
| BC066512.1 | - | 7 - 8 | 1216 | 70 | ||
| BC164728.1 | - | 7 - 8 | 949 | 70 | ||
More Information
Set Membership: |
|

stk25b
Genomic Map