Assay Details
| Gene Symbol: actr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213372.1 | NP_998537.1 | 10 - 11 | 1177 | 87 | |
| GenBank mRNA | BC046069.1 | - | 10 - 11 | 1177 | 87 | |
More Information
Set Membership: |
|

actr1
Genomic Map