Assay Details
| Gene Symbol: rap1ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243179.1 | NP_001230108.1 | 2 - 3 | 364 | 59 | |
| NM_213405.2 | NP_998570.2 | 3 - 4 | 462 | 59 | ||
| XM_021479589.1 | XP_021335264.1 | 1 - 2 | 1903 | 59 | ||
| XM_021479590.1 | XP_021335265.1 | 2 - 3 | 371 | 59 | ||
| GenBank mRNA | BC049510.1 | - | 3 - 4 | 430 | 59 | |
| EH543099.1 | - | 2 - 3 | 380 | 59 | ||
More Information
Set Membership: |
|

rap1ab
Genomic Map