Assay Details
| Gene Symbol: rap1ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243179.1 | NP_001230108.1 | 7 - 8 | 878 | 116 | |
| NM_213405.2 | NP_998570.2 | 8 - 9 | 976 | 116 | ||
| XM_021479589.1 | XP_021335264.1 | 6 - 7 | 2417 | 116 | ||
| GenBank mRNA | BC049510.1 | - | 8 - 9 | 944 | 116 | |
More Information
Set Membership: |
|

rap1ab
Genomic Map