Assay Details
| Gene Symbol: apex1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328578.1 | NP_001315507.1 | 3 - 4 | 264 | 117 | |
| NM_213421.2 | NP_998586.2 | 2 - 3 | 216 | 117 | ||
| XM_021474983.1 | XP_021330658.1 | 3 - 4 | 236 | 117 | ||
| GenBank mRNA | BC055545.1 | - | 2 - 3 | 117 | ||
| BC097053.1 | - | 2 - 3 | 117 | |||
| BC164240.1 | - | 1 - 2 | 117 | |||
| EF041101.1 | - | 2 - 3 | 117 | |||
| EF041102.1 | - | 2 - 3 | 117 | |||
More Information
Set Membership: |
|

apex1
Genomic Map