Assay Details
| Gene Symbol: prps1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001359894.1 | NP_001346823.1 | 4 - 5 | 715 | 134 | |
| NM_213327.2 | NP_998492.1 | 4 - 5 | 709 | 134 | ||
| NM_213533.3 | NP_998698.1 | 4 - 5 | 709 | 134 | ||
| GenBank mRNA | BC045301.1 | - | 4 - 5 | 134 | ||
| BC066485.1 | - | 4 - 5 | 693 | 134 | ||
| BC098599.1 | - | 4 - 5 | 714 | 134 | ||
| CT644358.2 | - | 3 - 4 | 391 | 134 | ||
More Information
Set Membership: |
|

prps1a
Genomic Map