Assay Details
| Gene Symbol: slc25a3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213722.1 | NP_998887.1 | 5 - 6 | 723 | 145 | |
| XM_005170654.3 | XP_005170711.1 | 5 - 6 | 145 | |||
| GenBank mRNA | BC046007.1 | - | 5 - 6 | 723 | 145 | |
| BC067565.1 | - | 5 - 6 | 681 | 145 | ||
More Information
Set Membership: |
|

slc25a3b
Genomic Map