Assay Details
| Gene Symbol: arrb2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214681.1 | NP_999846.1 | 11 - 12 | 1012 | 69 | |
| XM_009305269.3 | XP_009303544.1 | 11 - 12 | 1050 | 69 | ||
| GenBank mRNA | BC057519.1 | - | 11 - 12 | 1012 | 69 | |
| BC165790.1 | - | 11 - 12 | 952 | 69 | ||
More Information
Set Membership: |
|

arrb2a
Genomic Map