Assay Details
| Gene Symbol: arrb2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214681.1 | NP_999846.1 | 4 - 5 | 258 | 96 | |
| XM_009305269.3 | XP_009303544.1 | 4 - 5 | 296 | 96 | ||
| GenBank mRNA | BC057519.1 | - | 4 - 5 | 258 | 96 | |
| BC165790.1 | - | 4 - 5 | 198 | 96 | ||
More Information
Set Membership: |
|

arrb2a
Genomic Map