Assay Details
| Gene Symbol: st3gal3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214785.1 | NP_999950.1 | 5 - 6 | 432 | 155 | |
| XM_005163338.4 | XP_005163395.1 | 7 - 8 | 873 | 155 | ||
| XM_005163339.4 | XP_005163396.1 | 6 - 7 | 801 | 155 | ||
| XM_005163340.4 | XP_005163397.1 | 5 - 6 | 752 | 155 | ||
| XM_009298452.3 | XP_009296727.1 | 7 - 8 | 575 | 155 | ||
| XM_021479658.1 | XP_021335333.1 | 7 - 8 | 745 | 155 | ||
| XM_021479659.1 | XP_021335334.1 | 7 - 8 | 518 | 155 | ||
| GenBank mRNA | AJ626821.1 | - | 5 - 6 | 432 | 155 | |
| AJ705064.1 | - | 4 - 5 | 335 | 155 | ||
More Information
Set Membership: |
|

st3gal3b
Genomic Map