Assay Details
| Gene Symbol: st3gal3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214785.1 | NP_999950.1 | 10 - 11 | 1077 | 117 | |
| XM_005163338.4 | XP_005163395.1 | 12 - 13 | 1518 | 117 | ||
| XM_005163339.4 | XP_005163396.1 | 11 - 12 | 1446 | 117 | ||
| XM_005163340.4 | XP_005163397.1 | 10 - 11 | 1397 | 117 | ||
| XM_009298452.3 | XP_009296727.1 | 12 - 13 | 1220 | 117 | ||
| XM_021479658.1 | XP_021335333.1 | 12 - 13 | 1390 | 117 | ||
| XM_021479659.1 | XP_021335334.1 | 12 - 13 | 1163 | 117 | ||
| GenBank mRNA | AJ626821.1 | - | 10 - 11 | 1077 | 117 | |
| AJ705064.1 | - | 9 - 10 | 980 | 117 | ||
More Information
Set Membership: |
|

st3gal3b
Genomic Map