Assay Details
| Gene Symbol: sparc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001942.1 | NP_001001942.1 | 2 - 3 | 120 | 67 | |
| GenBank mRNA | AY575072.1 | - | 2 - 3 | 132 | 67 | |
| BC071436.1 | - | 2 - 3 | 120 | 67 | ||
| KM655589.1 | - | 1 - 2 | 53 | 67 | ||
More Information
Set Membership: |
|

sparc
Genomic Map