Assay Details
| Gene Symbol: med6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002084.1 | NP_001002084.1 | 7 - 8 | 649 | 63 | |
| GenBank mRNA | BC071547.1 | - | 7 - 8 | 649 | 63 | |
More Information
Set Membership: |
|

med6
Genomic Map