Assay Details
| Gene Symbol: ppp1r42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002202.1 | NP_001002202.1 | 9 - 10 | 1126 | 72 | |
| XM_009297386.3 | XP_009295661.1 | 8 - 9 | 1130 | 72 | ||
| XM_017353821.2 | XP_017209310.1 | 9 - 10 | 1237 | 72 | ||
| GenBank mRNA | BC072707.1 | - | 9 - 10 | 1126 | 72 | |
More Information
Set Membership: |
|

ppp1r42
Genomic Map