Assay Details
| Gene Symbol: prpf18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002204.1 | NP_001002204.1 | 6 - 7 | 686 | 74 | |
| XM_021467629.1 | XP_021323304.1 | 7 - 8 | 802 | 74 | ||
| GenBank mRNA | BC074081.1 | - | 6 - 7 | 686 | 74 | |
More Information
Set Membership: |
|

prpf18
Genomic Map