Assay Details
| Gene Symbol: tusc2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002402.2 | NP_001002402.1 | 2 - 3 | 356 | 71 | |
| GenBank mRNA | BC076181.1 | - | 2 - 3 | 357 | 71 | |
More Information
Set Membership: |
|

tusc2b
Genomic Map