Assay Details
| Gene Symbol: txn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002461.1 | NP_001002461.1 | 4 - 5 | 326 | 91 | |
| GenBank mRNA | BC076358.1 | - | 4 - 5 | 326 | 91 | |
| BC164445.1 | - | 4 - 5 | 300 | 91 | ||
More Information
Set Membership: |
|

txn
Genomic Map