Assay Details
| Gene Symbol: napgb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002469.2 | NP_001002469.1 | 10 - 11 | 836 | 91 | |
| GenBank mRNA | BC164093.1 | - | 10 - 11 | 697 | 91 | |
| EB871570.1 | - | 5 - 6 | 369 | 91 | ||
More Information
Set Membership: |
|

napgb
Genomic Map