Assay Details
| Gene Symbol: slc1a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002513.2 | NP_001002513.2 | 4 - 5 | 961 | 62 | |
| XM_005156786.3 | XP_005156843.1 | 5 - 6 | 1235 | 62 | ||
| GenBank mRNA | HM138695.1 | - | 5 - 6 | 1041 | 62 | |
More Information
Set Membership: |
|

slc1a4
Genomic Map