Assay Details
| Gene Symbol: celf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002562.2 | NP_001002562.1 | 13 - 14 | 1929 | 142 | |
| XM_009295191.3 | XP_009293466.1 | 13 - 14 | 2097 | 142 | ||
| XM_009295192.3 | XP_009293467.1 | 13 - 14 | 2094 | 142 | ||
| XM_009295193.3 | XP_009293468.1 | 13 - 14 | 2099 | 142 | ||
| XM_009295194.3 | XP_009293469.1 | 13 - 14 | 2039 | 142 | ||
| XM_017352768.2 | XP_017208257.1 | 13 - 14 | 2028 | 142 | ||
| XM_021468805.1 | XP_021324480.1 | 12 - 13 | 1919 | 142 | ||
| XM_021468806.1 | XP_021324481.1 | 12 - 13 | 1915 | 142 | ||
| XM_021468807.1 | XP_021324482.1 | 12 - 13 | 1861 | 142 | ||
| XM_021468808.1 | XP_021324483.1 | 12 - 13 | 1920 | 142 | ||
| GenBank mRNA | BC076238.1 | - | 13 - 14 | 1931 | 142 | |
More Information
Set Membership: |
|

celf4
Genomic Map