Assay Details
| Gene Symbol: pmepa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002580.1 | NP_001002580.1 | 334 | 69 | ||
| GenBank mRNA | BC076211.1 | - | 334 | 69 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pmepa1| Gene Symbol: pmepa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002580.1 | NP_001002580.1 | 334 | 69 | ||
| GenBank mRNA | BC076211.1 | - | 334 | 69 | ||
Set Membership: |
|