Assay Details
| Gene Symbol: supt3h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002625.1 | NP_001002625.1 | 2 - 3 | 179 | 66 | |
| XM_021467143.1 | XP_021322818.1 | 2 - 3 | 210 | 66 | ||
| GenBank mRNA | BC075959.1 | - | 2 - 3 | 179 | 66 | |
More Information
Set Membership: |
|

supt3h
Genomic Map