Assay Details
| Gene Symbol: rnaseh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002659.2 | NP_001002659.2 | 3 - 4 | 615 | 71 | |
| NM_001285828.1 | NP_001272757.1 | 4 - 5 | 433 | 71 | ||
| GenBank mRNA | BC076457.1 | - | 3 - 4 | 596 | 71 | |
| BC164625.1 | - | 3 - 4 | 411 | 71 | ||
| EH542009.1 | - | 3 - 4 | 654 | 71 | ||
| EH544014.1 | - | 4 - 5 | 456 | 71 | ||
More Information
Set Membership: |
|

rnaseh1
Genomic Map