Assay Details
| Gene Symbol: rcan1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003616.1 | NP_001003616.1 | 1 - 2 | 397 | 83 | |
| GenBank mRNA | BC078255.1 | - | 1 - 2 | 397 | 83 | |
More Information
Set Membership: |
|

rcan1a
Genomic Map