Assay Details
| Gene Symbol: acads | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003743.1 | NP_001003743.1 | 7 - 8 | 915 | 65 | |
| GenBank mRNA | BC079521.1 | - | 7 - 8 | 915 | 65 | |
More Information
Set Membership: |
|

acads
Genomic Map