Assay Details
| Gene Symbol: chst10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003779.1 | NP_001003779.1 | 5 - 6 | 775 | 89 | |
| XM_005159888.4 | XP_005159945.1 | 5 - 6 | 662 | 89 | ||
| XM_005159889.4 | XP_005159946.1 | 5 - 6 | 659 | 89 | ||
| XM_005159890.4 | XP_005159947.1 | 5 - 6 | 782 | 89 | ||
| XM_005159891.4 | XP_005159948.1 | 5 - 6 | 779 | 89 | ||
| XM_005159892.4 | XP_005159949.1 | 5 - 6 | 804 | 89 | ||
| GenBank mRNA | BC079482.1 | - | 5 - 6 | 775 | 89 | |
More Information
Set Membership: |
|

chst10
Genomic Map