Assay Details
| Gene Symbol: sept5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003782.2 | NP_001003782.1 | 5 - 6 | 377 | 60 | |
| XM_021479157.1 | XP_021334832.1 | 5 - 6 | 442 | 60 | ||
| XM_021479158.1 | XP_021334833.1 | 4 - 5 | 355 | 60 | ||
| GenBank mRNA | BC078256.1 | - | 5 - 6 | 381 | 60 | |
More Information
Set Membership: |
|

sept5b
Genomic Map