Assay Details
| Gene Symbol: tap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006594.1 | NP_001006594.1 | 3 - 4 | 742 | 81 | |
| GenBank mRNA | BC163447.1 | - | 3 - 4 | 776 | 81 | |
| BC163453.1 | - | 3 - 4 | 776 | 81 | ||
More Information
Set Membership: |
|

tap2a
Genomic Map