Assay Details
| Gene Symbol: tap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006594.1 | NP_001006594.1 | 7 - 8 | 1464 | 92 | |
| GenBank mRNA | BC163447.1 | - | 7 - 8 | 1498 | 92 | |
| BC163453.1 | - | 7 - 8 | 1498 | 92 | ||
More Information
Set Membership: |
|

tap2a
Genomic Map