Assay Details
| Gene Symbol: hapln1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007790.2 | NP_001007791.2 | 1 - 2 | 221 | 67 | |
| NM_001313730.1 | NP_001300659.1 | 1 - 2 | 221 | 67 | ||
| GenBank mRNA | AB167511.1 | - | 1 - 2 | 57 | 67 | |
| BC142597.1 | - | 1 - 2 | 217 | 67 | ||
| BC163127.1 | - | 1 - 2 | 117 | 67 | ||
| EH448450.1 | - | 1 - 2 | 249 | 67 | ||
| KM655692.1 | - | 1 - 2 | 57 | 67 | ||
More Information
Set Membership: |
|

hapln1a
Genomic Map