Assay Details
| Gene Symbol: atp6v1e1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001365440.1 | NP_001352369.1 | 1 - 2 | 217 | 103 | |
| GenBank mRNA | BC086733.1 | - | 1 - 2 | 217 | 103 | |
| DT073384.1 | - | 1 - 2 | 184 | 103 | ||
| EH591399.1 | - | 1 - 2 | 224 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC086733.1, NM_001365440.1 transcripts.
Set Membership: |
|

atp6v1e1a
Genomic Map