Assay Details
| Gene Symbol: atp6v1e1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001365440.1 | NP_001352369.1 | 3 - 4 | 396 | 112 | |
| GenBank mRNA | BC086733.1 | - | 3 - 4 | 396 | 112 | |
| BC165161.1 | - | 3 - 4 | 244 | 112 | ||
| EH591399.1 | - | 3 - 4 | 403 | 112 | ||
More Information
Set Membership: |
|

atp6v1e1a
Genomic Map