Assay Details
| Gene Symbol: ptgr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008651.1 | NP_001008651.1 | 4 - 5 | 292 | 71 | |
| GenBank mRNA | BC086722.1 | - | 4 - 5 | 292 | 71 | |
More Information
Set Membership: |
|

ptgr1
Genomic Map