Assay Details
| Gene Symbol: ch25h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008652.1 | NP_001008652.1 | 1 | 652 | 67 | |
| GenBank mRNA | BC086721.1 | - | 1 | 652 | 67 | |
| BC165116.1 | - | 1 | 656 | 67 | ||
| MF095414.1 | - | 1 | 622 | 67 | ||
More Information
Set Membership: |
|

ch25h
Genomic Map