Assay Details
| Gene Symbol: xkr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012253.2 | NP_001012253.2 | 2 - 3 | 485 | 62 | |
| GenBank mRNA | AY534279.1 | - | 2 - 3 | 472 | 62 | |
| BC107954.1 | - | 2 - 3 | 485 | 62 | ||
More Information
Set Membership: |
|

xkr7
Genomic Map