Assay Details
| Gene Symbol: adcyap1r1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013444.2 | NP_001013462.1 | 1384 | 121 | ||
| NM_001142925.1 | NP_001136397.1 | 1300 | 121 | |||
| NM_001142926.1 | NP_001136398.1 | 1381 | 121 | |||
| GenBank mRNA | AY738798.2 | - | 1300 | 121 | ||
| AY738800.2 | - | 1381 | 121 | |||
| AY738801.2 | - | 1406 | 121 | |||
| AY738802.2 | - | 1384 | 121 | |||
More Information
Set Membership: |
|

adcyap1r1a
Genomic Map