Assay Details
| Gene Symbol: eif4g2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014289.2 | NP_001014311.1 | 13 - 14 | 1596 | 125 | |
| GenBank mRNA | AB096100.1 | - | 13 - 14 | 1662 | 125 | |
| BC059195.1 | - | 13 - 14 | 1595 | 125 | ||
| BC165852.1 | - | 6 - 7 | 791 | 125 | ||
More Information
Set Membership: |
|

eif4g2a
Genomic Map