Assay Details
| Gene Symbol: impdh1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014369.2 | NP_001014391.1 | 11 - 12 | 1393 | 61 | |
| XM_005164717.4 | XP_005164774.1 | 12 - 13 | 1899 | 61 | ||
| XM_005164718.4 | XP_005164775.1 | 11 - 12 | 1821 | 61 | ||
| XM_017354952.2 | XP_017210441.1 | 12 - 13 | 1489 | 61 | ||
| XM_017354953.2 | XP_017210442.1 | 6 - 7 | 755 | 61 | ||
| GenBank mRNA | BC091790.1 | - | 11 - 12 | 1315 | 61 | |
| CT657866.2 | - | 5 - 6 | 552 | 61 | ||
| DN896992.1 | - | 3 - 4 | 338 | 61 | ||
More Information
Set Membership: |
|

impdh1b
Genomic Map