Assay Details
| Gene Symbol: ets1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017558.1 | NP_001017558.1 | 4 - 5 | 625 | 68 | |
| GenBank mRNA | BC092935.1 | - | 4 - 5 | 625 | 68 | |
| BC165216.1 | - | 484 | 68 | |||
| KF774190.1 | - | 4 - 5 | 578 | 68 | ||
More Information
Set Membership: |
|

ets1
Genomic Map