Assay Details
| Gene Symbol: camk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017607.1 | NP_001017607.1 | 5 - 6 | 398 | 63 | |
| GenBank mRNA | BC092841.1 | - | 5 - 6 | 398 | 63 | |
| BC164407.1 | - | 4 - 5 | 364 | 63 | ||
More Information
Set Membership: |
|

camk4
Genomic Map