Assay Details
| Gene Symbol: zbtb8os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017687.1 | NP_001017687.1 | 6 - 7 | 432 | 112 | |
| GenBank mRNA | BC093322.1 | - | 6 - 7 | 432 | 112 | |
More Information
Set Membership: |
|

zbtb8os
Genomic Map