Assay Details
| Gene Symbol: adtrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017719.1 | NP_001017719.1 | 4 - 5 | 548 | 127 | |
| GenBank mRNA | BC093244.1 | - | 4 - 5 | 548 | 127 | |
More Information
Set Membership: |
|

adtrp1
Genomic Map