Assay Details
| Gene Symbol: mecr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017852.1 | NP_001017852.1 | 3 - 4 | 544 | 72 | |
| XM_021466687.1 | XP_021322362.1 | 1 - 2 | 242 | 72 | ||
| XM_021466688.1 | XP_021322363.1 | 3 - 4 | 528 | 72 | ||
| GenBank mRNA | BC072704.1 | - | 3 - 4 | 524 | 72 | |
| BC092759.1 | - | 3 - 4 | 544 | 72 | ||
More Information
Set Membership: |
|

mecr
Genomic Map