Assay Details
| Gene Symbol: slc2a15b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020494.1 | NP_001018330.1 | 3 - 4 | 415 | 71 | |
| GenBank mRNA | BC094298.1 | - | 3 - 4 | 415 | 71 | |
| BC165759.1 | - | 3 - 4 | 334 | 71 | ||
More Information
Set Membership: |
|

slc2a15b
Genomic Map